5-Brom-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
FM-2496-1G
FM-2496-250M
COBBFM-2496-1GEA
497
EUR
COBBFM-2496-1G
COBBFM-2496-250M
5-Brom-2-indolboronsäurepinakolester
5-Brom-2-indolboronsäurepinakolester
Formel:
C₁₄H₁₇BBrNO₂ Molecular Weight: 322,0053 g/mol |
MDL:
MFCD11858360 CAS-Nummer: 1256358-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...