6-Brom-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
FM-2511-1G
FM-2511-250M
COBBFM-2511-1GEA
1010
EUR
COBBFM-2511-1G
COBBFM-2511-250M
6-Brom-2-indolboronsäurepinakolester
6-Brom-2-indolboronsäurepinakolester
Formel:
C₁₄H₁₇BBrNO₂ Molecular Weight: 322,01 g/mol |
MDL:
MFCD11858393 CAS-Nummer: 1218791-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...