1-Boc-6-brom-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 6-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6223-1G PN-6223-250M
COBBPN-6223-1GEA 1430 EUR
COBBPN-6223-1G COBBPN-6223-250M
1-Boc-6-brom-3-indolboronsäurepinakolester
1-Boc-6-brom-3-indolboronsäurepinakolester
Formel: C₁₉H₂₅BBrNO₄
Molecular Weight: 422,1200 g/mol
MDL: MFCD15143608
CAS-Nummer: 1218790-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR