5-Chlor-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
FM-2495-5G
FM-2495-1G
COBBFM-2495-5GEA
2510
EUR
COBBFM-2495-5G
COBBFM-2495-1G
5-Chlor-2-indolboronsäurepinakolester
5-Chlor-2-indolboronsäurepinakolester
Formel:
C₁₄H₁₇BClNO₂ Molecular Weight: 277.5543 g/mol |
MDL:
MFCD11858358 CAS-Nummer: 1256358-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...