6-Chlor-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-6267-1G PN-6267-250M
COBBPN-6267-1GEA 787 EUR
COBBPN-6267-1G COBBPN-6267-250M
6-Chlor-2-indolboronsäurepinakolester
6-Chlor-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BClNO₂
Molecular Weight: 277.5543 g/mol
MDL: MFCD16295112
CAS-Nummer: 912331-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR