7-Chlor-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 7-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-6268-1G PN-6268-250M
COBBPN-6268-1GEA 1240 EUR
COBBPN-6268-1G COBBPN-6268-250M
7-Chlor-2-indolboronsäurepinakolester
7-Chlor-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BClNO₂
Molecular Weight: 277,55 g/mol
MDL: MFCD16295113
CAS-Nummer: 936901-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR