1-Boc-5-chlor-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6252-1G PN-6252-250M
COBBPN-6252-1GEA 679 EUR
COBBPN-6252-1G COBBPN-6252-250M
1-Boc-5-chlor-3-indolboronsäurepinakolester
1-Boc-5-chlor-3-indolboronsäurepinakolester
Formel: C₁₉H₂₅BClNO₄
Molecular Weight: 377.6701 g/mol
MDL: MFCD15143612
CAS-Nummer: 1218790-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR