1-Boc-7-chlor-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 7-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6253-1G PN-6253-250M
COBBPN-6253-1GEA 742 EUR
COBBPN-6253-1G COBBPN-6253-250M
1-Boc-7-chlor-3-indolboronsäurepinakolester
1-Boc-7-chlor-3-indolboronsäurepinakolester
Formel: C₁₉H₂₅BClNO₄
Molecular Weight: 377,67 g/mol
MDL: MFCD15143613
CAS-Nummer: 1218790-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR