4-Fluor-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
PN-6269-1G
PN-6269-250M
COBBPN-6269-1GEA
623
EUR
COBBPN-6269-1G
COBBPN-6269-250M
4-Fluor-2-indolboronsäurepinakolester
4-Fluor-2-indolboronsäurepinakolester
Formel:
C₁₄H₁₇BFNO₂ Molecular Weight: 261,0997 g/mol |
MDL:
MFCD16618972 CAS-Nummer: 1256359-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...