5-Fluor-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2498-5G FM-2498-1G
COBBFM-2498-5GEA 2460 EUR
COBBFM-2498-5G COBBFM-2498-1G
5-Fluor-2-indolboronsäurepinakolester
5-Fluor-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BFNO₂
Molecular Weight: 261.0997 g/mol
MDL: MFCD11858362
CAS-Nummer: 1256358-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR