1-Methyl-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-2401-5G PN-2401-1G
COBBPN-2401-5GEA 560 EUR
COBBPN-2401-5G COBBPN-2401-1G
1-Methyl-2-indolboronsäurepinakolester
1-Methyl-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₂
Molecular Weight: 257,135 g/mol
MDL: MFCD13182063
CAS-Nummer: 596819-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR