1-Methyl-4-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
PN-9107-1G
PN-9107-250M
COBBPN-9107-1GEA
249
EUR
COBBPN-9107-1G
COBBPN-9107-250M
1-Methyl-4-indolboronsäurepinakolester
1-Methyl-4-indolboronsäurepinakolester
Formel:
C₁₅H₂₀BNO₂ Molecular Weight: 257,1400 g/mol |
MDL:
MFCD08690266 CAS-Nummer: 898289-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...