4-Methyl-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2512-1G FM-2512-250M
COBBFM-2512-1GEA 822 EUR
COBBFM-2512-1G COBBFM-2512-250M
4-Methyl-2-indolboronsäurepinakolester
4-Methyl-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₂
Molecular Weight: 257.1358 g/mol
MDL: MFCD11858397
CAS-Nummer: 1072811-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR