6-Methyl-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
FM-2505-1G
FM-2505-250M
COBBFM-2505-1GEA
593
EUR
COBBFM-2505-1G
COBBFM-2505-250M
6-Methyl-2-indolboronsäurepinakolester
6-Methyl-2-indolboronsäurepinakolester
Formel:
C₁₅H₂₀BNO₂ Molecular Weight: 257.1358 g/mol |
MDL:
MFCD11858378 CAS-Nummer: 912332-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...