7-Methyl-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 7-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2503-1G FM-2503-250M
COBBFM-2503-1GEA 865 EUR
COBBFM-2503-1G COBBFM-2503-250M
7-Methyl-2-indolboronsäurepinakolester
7-Methyl-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₂
Molecular Weight: 257.1358 g/mol
MDL: MFCD11858374
CAS-Nummer: 912331-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR