6-Chlor-1-methyl-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-5107-1G PN-5107-5G
COBBPN-5107-1GEA 623 EUR
COBBPN-5107-1G COBBPN-5107-5G
6-Chlor-1-methyl-2-indolboronsäurepinakolester
6-Chlor-1-methyl-2-indolboronsäurepinakolester
Formel: C₁₅H₁₉BClNO₂
Molecular Weight: 291.58 g/mol
MDL: MFCD11504964
CAS-Nummer: 1073353-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR