6-Chlor-1-methyl-2-indolboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-Chloro-1-methylindole-2-boronic acid
BB-5107-1G
BB-5107-250M
COBBBB-5107-1GEA
742
EUR
COBBBB-5107-1G
COBBBB-5107-250M
6-Chlor-1-methyl-2-indolboronsäure
6-Chloro-1-methyl-2-indoleboronic acid
Formel:
C₉H₉BClNO₂ Molecular Weight: 209.44 g/mol |
MDL:
MFCD09878352 CAS-Nummer: 957066-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...