5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd

Lieferant: FLUOROCHEM

CFF490.5 CFF490.25 017498-5G 017498-25G 017498-1G
CHMPCFF490.5EA 321 EUR
CHMPCFF490.5 CHMPCFF490.25 FLUO017498-5G FLUO017498-25G FLUO017498-1G
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
Formel: C₉H₁₁ClN₂O
Molecular Weight: 158,59 g/mol
MDL: MFCD00052538
CAS-Nummer: 27006-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR