5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
Lieferant: Thermo Scientific
EN00019.1
EN00019EA
MAYBEN00019.1EA
0
EUR
MAYBEN00019.1
MAYBEN00019EA
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
Formel:
C₉H₁₁ClN₂O Molecular Weight: 158,59 g/mol |
MDL:
MFCD00052538 CAS-Nummer: 27006-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...