5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonylchlorid

Lieferant: FLUOROCHEM

CFF416.5 CFF416.10 CFF416.25 017438-5G 017438-10G 017438-25G 017438-1G
CHMPCFF416.5EA 348 EUR
CHMPCFF416.5 CHMPCFF416.10 CHMPCFF416.25 FLUO017438-5G FLUO017438-10G FLUO017438-25G FLUO017438-1G
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonylchlorid
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonylchlorid
Formel: C₅H₆Cl₂N₂O₂S
MDL: MFCD00052536
CAS-Nummer: 88398-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR