5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonylchlorid
Lieferant: Thermo Scientific
CD10088DA
CD10088EA
MAYBCD10088DAEA
0
EUR
MAYBCD10088DA
MAYBCD10088EA
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonylchlorid
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonylchlorid
Formel:
C₅H₆Cl₂N₂O₂S |
MDL:
MFCD00052536 CAS-Nummer: 88398-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...