4-Brom-1-(2-chlorphenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1355-5G HI-1355-1G HI-1355-25G
COBBHI-1355-5GEA 451 EUR
COBBHI-1355-5G COBBHI-1355-1G COBBHI-1355-25G
4-Brom-1-(2-chlorphenyl)-1H-pyrazol
4-Bromo-1-(2-chlorophenyl)-1H-pyrazole
Formel: C₉H₆BrClN₂
MDL: MFCD09878407
CAS-Nummer: 957063-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR