4-Brom-1-(3-chlorphenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1315-5G HI-1315-1G
COBBHI-1315-5GEA 451 EUR
COBBHI-1315-5G COBBHI-1315-1G
4-Brom-1-(3-chlorphenyl)-1H-pyrazol
4-Bromo-1-(3-chlorophenyl)-1H-pyrazole
Formel: C₉H₆BrClN₂
Molecular Weight: 257,5100 g/mol
MDL: MFCD09475849
CAS-Nummer: 957034-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR