4-Brom-1-(4-fluorphenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1332-1G HI-1332-25G HI-1332-5G
COBBHI-1332-1GEA 149 EUR
COBBHI-1332-1G COBBHI-1332-25G COBBHI-1332-5G
4-Brom-1-(4-fluorphenyl)-1H-pyrazol
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole
Formel: C₉H₆BrFN₂
Molecular Weight: 241,0600 g/mol
MDL: MFCD09801028
CAS-Nummer: 957062-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR