Ethyl-5-amino-4-cyan-1-(3-methoxyphenyl)-1H-pyrazol-3-carboxylat
Lieferant: COMBI-BLOCKS
HC-2395-1G
HC-2395-250M
COBBHC-2395-1GEA
141
EUR
COBBHC-2395-1G
COBBHC-2395-250M
Ethyl-5-amino-4-cyan-1-(3-methoxyphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(3-methoxyphenyl)-1H-pyrazol-3-carboxylat
Formel:
C₁₄H₁₄N₄O₃ |
MDL:
MFCD09835750 CAS-Nummer: 1150164-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...