Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat

Lieferant: Thermo Scientific

Synonyme: Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate

RH00119DA RH00119EA
MAYBRH00119DAEA 0 EUR
MAYBRH00119DA MAYBRH00119EA
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Formel: C₁₂H₁₂FN₃O₂
MDL: MFCD00173917
CAS-Nummer: 138907-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR