3,6-Dimethoxy-4-pyridazinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,6-Dimethoxylpyridazine-4-boronic acid

BB-9008-1G BB-9008-5G
COBBBB-9008-1GEA 149 EUR
COBBBB-9008-1G COBBBB-9008-5G
3,6-Dimethoxy-4-pyridazinboronsäure
3,6-Dimethoxypyridazine-4-boronic acid
Formel: C₆H₉BN₂O₄
MDL: MFCD10700157
CAS-Nummer: 1015480-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR