4,5-Dibrom-3(2H)-pyridazinon

Lieferant: BIONET RESEARCH

Synonyme: 4,5-Dibromopyridazin-3(2H)-one

7M-627S.500 7M-627S.1 7M-627S.5 7M-627S.10
BIOT7M-627S.500EA 0 EUR
BIOT7M-627S.500 BIOT7M-627S.1 BIOT7M-627S.5 BIOT7M-627S.10
4,5-Dibrom-3(2H)-pyridazinon
4,5-Dibrom-3(2H)-pyridazinon
Formel: C₄H₂Br₂N₂O
Molecular Weight: 253.89 g/mol
MDL: MFCD00023641
CAS-Nummer: 5788-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR