3-Chlor-6-(ethylamino)pyridazin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-N-ethylpyridazin-3-amine

HC-6093-1G HC-6093-5G
COBBHC-6093-1GEA 149 EUR
COBBHC-6093-1G COBBHC-6093-5G
3-Chlor-6-(ethylamino)pyridazin
3-Chloro-6-(ethylamino)pyridazine
Formel: C₆H₈ClN₃
Molecular Weight: 157,6008 g/mol
MDL: MFCD08363097
CAS-Nummer: 68588-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR