3-Chlor-6-(methylamino)pyridazin
Lieferant: COMBI-BLOCKS
HC-6177-5G
HC-6177-1G
COBBHC-6177-5GEA
77.4
EUR
COBBHC-6177-5G
COBBHC-6177-1G
3-Chlor-6-(methylamino)pyridazin
3-Chloro-6-(methylamino)pyridazine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,5700 g/mol |
MDL:
MFCD02178601 CAS-Nummer: 14959-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...