3-Chlor-6-(pentylamino)pyridazin

Lieferant: COMBI-BLOCKS

HC-6138-5G HC-6138-1G
COBBHC-6138-5GEA 303 EUR
COBBHC-6138-5G COBBHC-6138-1G
3-Chlor-6-(pentylamino)pyridazin
3-Chloro-6-(pentylamino)pyridazine
Formel: C₉H₁₄ClN₃
MDL: MFCD09800954
CAS-Nummer: 941294-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR