3-Chlor-6-(3-methoxypropylamino)pyridazin

Lieferant: COMBI-BLOCKS

HC-6185-5G HC-6185-1G
COBBHC-6185-5GEA 375 EUR
COBBHC-6185-5G COBBHC-6185-1G
3-Chlor-6-(3-methoxypropylamino)pyridazin
3-Chlor-6-(3-methoxypropylamino)pyridazin
Formel: C₈H₁₂ClN₃O
MDL: MFCD08363096
CAS-Nummer: 898656-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR