3-Chlor-6-(cyclohexylamino)pyridazin

Lieferant: COMBI-BLOCKS

HC-6188-5G HC-6188-1G
COBBHC-6188-5GEA 595 EUR
COBBHC-6188-5G COBBHC-6188-1G
3-Chlor-6-(cyclohexylamino)pyridazin
3-Chloro-6-(cyclohexylamino)pyridazine
Formel: C₁₀H₁₄ClN₃
MDL: MFCD00968568
CAS-Nummer: 1014-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR