3-Chlor-6-(cyclopropylamino)pyridazin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-N-cyclopropylpyridazin-3-amine

HC-6145-5G HC-6145-1G
COBBHC-6145-5GEA 399 EUR
COBBHC-6145-5G COBBHC-6145-1G
3-Chlor-6-(cyclopropylamino)pyridazin
3-Chloro-6-(cyclopropylamino)pyridazine
Formel: C₇H₈ClN₃
Molecular Weight: 169.6100 g/mol
MDL: MFCD09800959
CAS-Nummer: 941294-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR