4-Pyridazincarbonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Pyridazinecarboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
297763-1G
297763-5G
SIAL297763-1GEA
56.2
EUR
SIAL297763-1G
SIAL297763-5G
4-Pyridazincarbonsäure, Sigma-Aldrich®
Pyridazine-4-carboxylic acid
Formel:
C₅H₄N₂O₂ Molecular Weight: 124,10 g/mol |
MDL:
MFCD00011576 CAS-Nummer: 50681-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...