6-Methyl-3(2H)-pyridazinon ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Methylpyridazin-3(2H)-one
A14471.06
A14471.14
A14471.06EA
104
EUR
A14471.06
A14471.14
6-Methyl-3(2H)-pyridazinon ≥98%
6-Methyl-3(2H)-pyridazinon
Purity: 98%
CAS No.: 13327-27-0
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Melting Point: 143-145°
Beilstein Reference: 109928
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Schmelzpunkt: 143…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039720 CAS-Nummer: 13327-27-0 EINECS: 236-367-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...