6-Methyl-3(2H)-pyridazinon

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methylpyridazin-3(2H)-one

PY-1366-5G PY-1366-1G
COBBPY-1366-5GEA 38.2 EUR
COBBPY-1366-5G COBBPY-1366-1G
6-Methyl-3(2H)-pyridazinon
6-Methyl-3(2H)-pyridazinon
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
MDL: MFCD00039720
CAS-Nummer: 13327-27-0
EINECS: 236-367-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR