3-(Cyclopropylmethoxy)-5-methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-(Cyclopropylmethoxy)-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5984-1G PN-5984-250M
COBBPN-5984-1GEA 303 EUR
COBBPN-5984-1G COBBPN-5984-250M
3-(Cyclopropylmethoxy)-5-methylphenylboronsäurepinakolester
3-(Cyclopropylmethoxy)-5-methylphenylboronsäurepinakolester
Formel: C₁₇H₂₅BO₃
Molecular Weight: 288.1896 g/mol
MDL: MFCD12546604
CAS-Nummer: 1218789-80-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR