6-Chlor-2,5-dimethylbenzimidazol

Lieferant: COMBI-BLOCKS

HC-3318-5G HC-3318-1G
COBBHC-3318-5GEA 227 EUR
COBBHC-3318-5G COBBHC-3318-1G
6-Chlor-2,5-dimethylbenzimidazol
6-Chlor-2,5-dimethylbenzimidazol
Formel: C₉H₉ClN₂
MDL: MFCD15143570
CAS-Nummer: 221548-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR