6-Nitrobenzoxazol

Lieferant: BIONET RESEARCH

Synonyme: 6-Nitrobenzo[d]oxazole

2P-029.500 2P-029.1 2P-029.5 2P-029.10
BIOT2P-029.500EA 0 EUR
BIOT2P-029.500 BIOT2P-029.1 BIOT2P-029.5 BIOT2P-029.10
6-Nitrobenzoxazol
6-Nitrobenzoxazole
Formel: C₇H₄N₂O₃
Molecular Weight: 164.1183 g/mol
MDL: MFCD00797933
CAS-Nummer: 17200-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR