5-Brom-2-methylbenzoxazol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-methyl-1,3-benzoxazole

HI-1572-5G HI-1572-1G
COBBHI-1572-5GEA 259 EUR
COBBHI-1572-5G COBBHI-1572-1G
5-Brom-2-methylbenzoxazol
5-Bromo-2-methylbenzoxazole
Formel: C₈H₆BrNO
Molecular Weight: 212,0400 g/mol
MDL: MFCD08702771
CAS-Nummer: 5676-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR