1H-Indazol-6-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
PN-0147-5G
PN-0147-1G
COBBPN-0147-5GEA
449
EUR
COBBPN-0147-5G
COBBPN-0147-1G
1H-Indazol-6-boronsäurepinakolester
1H-Indazol-6-boronsäurepinakolester
Formel:
C₁₃H₁₇BN₂O₂ Molecular Weight: 244,0973 g/mol |
MDL:
MFCD07368067 CAS-Nummer: 937049-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...