1H-Indazol-4-boronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-9063-1G BB-9063-250M
COBBBB-9063-1GEA 286 EUR
COBBBB-9063-1G COBBBB-9063-250M
1H-Indazol-4-boronsäure Hydrochlorid
1H-Indazol-4-boronsäure Hydrochlorid
Formel: C₇H₈BClN₂O₂
MDL: MFCD11054042
CAS-Nummer: 1252598-02-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR