7-Brom-5-chlor-1H-indazol

Lieferant: COMBI-BLOCKS

IN-0143-5G IN-0143-1G
COBBIN-0143-5GEA 421 EUR
COBBIN-0143-5G COBBIN-0143-1G
7-Brom-5-chlor-1H-indazol
7-Bromo-5-chloro-1H-indazole
Formel: C₇H₄BrClN₂
MDL: MFCD09258643
CAS-Nummer: 875305-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR