3-Amino-5-brom-1H-indazol
Lieferant: BIONET RESEARCH
Synonyme:
5-Bromo-1H-indazol-3-amine
10Y-0709.1
10Y-0709.10
10Y-0709.5
10Y-0709.500
BIOT10Y-0709.1EA
0
EUR
BIOT10Y-0709.1
BIOT10Y-0709.10
BIOT10Y-0709.5
BIOT10Y-0709.500
3-Amino-5-brom-1H-indazol
3-Amino-5-bromo-1H-indazole
Formel:
C₇H₆BrN₃ Molecular Weight: 212.05 g/mol |
MDL:
MFCD03426696 CAS-Nummer: 61272-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...