3-Amino-4-chlor-1H-indazol

Lieferant: BIONET RESEARCH

Synonyme: 4-Chloro-1H-indazol-3-amine

10Y-0704.1 10Y-0704.500 10Y-0704.10 10Y-0704.5
BIOT10Y-0704.1EA 0 EUR
BIOT10Y-0704.1 BIOT10Y-0704.500 BIOT10Y-0704.10 BIOT10Y-0704.5
3-Amino-4-chlor-1H-indazol
3-Amino-4-chloro-1H-indazole
Formel: C₇H₆ClN₃
Molecular Weight: 167,60 g/mol
MDL: MFCD00663139
CAS-Nummer: 20925-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR