Methyl-1H-indazol-4-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 1H-indazole-4-carboxylate
HC-3022-1G
HC-3022-5G
COBBHC-3022-1GEA
48.6
EUR
COBBHC-3022-1G
COBBHC-3022-5G
Methyl-1H-indazol-4-carboxylat
Methyl-1H-indazol-4-carboxylat
Formel:
C₉H₈N₂O₂ Molecular Weight: 176.1700 g/mol |
MDL:
MFCD07371610 CAS-Nummer: 192945-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...