Methyl-1H-indazol-5-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 1H-indazole-5-carboxylate

HC-2722-5G HC-2722-1G
COBBHC-2722-5GEA 76.3 EUR
COBBHC-2722-5G COBBHC-2722-1G
Methyl-1H-indazol-5-carboxylat
Methyl-1H-indazol-5-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1700 g/mol
MDL: MFCD07371609
CAS-Nummer: 473416-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR