1H-Indazol-3-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indazole-3-carboxylic acid

56915-10G 56915-50G
SIAL56915-10GEA 99.6 EUR
SIAL56915-10G SIAL56915-50G
1H-Indazol-3-carbonsäure, Sigma-Aldrich®
1H-Indazole-3-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD00211066
CAS-Nummer: 4498-67-3
EINECS: 224-794-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR