6-Iod-1H-indazol

Lieferant: CHEMPUR

Synonyme: 6-Iodoindazole

45-0085.1 45-0085.5
CHMP45-0085.1EA 0 EUR
CHMP45-0085.1 CHMP45-0085.5
6-Iod-1H-indazol
6-Iodo-1H-indazole
Formel: C₇H₅IN₂
Molecular Weight: 244,0300 g/mol
MDL: MFCD04114695
CAS-Nummer: 261953-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR